SY3H3D680FAC Synthonix Corporation

7-Methoxy-4-oxo-1,4-dihydroquinoline-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 77474-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/55402.html

Safety Information

MSDS https://synthonix.com/55402.html
Availability details for
Quantity
  • Availability
  • Add to Cart