SY3H3D680FAE Synthonix Corporation

Methyl 7-bromo-3-oxo-2,3-dihydro-1H-indene-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 1951439-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55410.html

Safety Information

MSDS https://synthonix.com/55410.html
Availability details for
Quantity
  • Availability
  • Add to Cart