SY3H3D680FB5 Synthonix Corporation

Methyl 7-bromo-1H-indole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 885523-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/53749.html

Safety Information

MSDS https://synthonix.com/53749.html
Availability details for
Quantity
  • Availability
  • Add to Cart