SY3H3D680FB6 Synthonix Corporation

Methyl 4-amino-3-cyclopropylbenzoate hydrochloride

  • Cas Number : 2044702-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55461.html

Safety Information

MSDS https://synthonix.com/55461.html
Availability details for
Quantity
  • Availability
  • Add to Cart