SY3H3D680FBA Synthonix Corporation

(R)-Methyl 3-amino-2,3-dihydro-1H-indene-5-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1246509-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55466.html

Safety Information

MSDS https://synthonix.com/55466.html
Availability details for
Quantity
  • Availability
  • Add to Cart