SY3H3D680FC7 Synthonix Corporation

(R)-6-Methoxy-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 911372-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55502.html

Safety Information

MSDS https://synthonix.com/55502.html
Availability details for
Quantity
  • Availability
  • Add to Cart