SY3H3D680FC8 Synthonix Corporation

(S)-6-Methoxy-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 911372-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55503.html

Safety Information

MSDS https://synthonix.com/55503.html
Availability details for
Quantity
  • Availability
  • Add to Cart