SY3H3D680FC9 Synthonix Corporation

3-(4-Methoxyphenyl)morpholine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1171560-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55504.html

Safety Information

MSDS https://synthonix.com/55504.html
Availability details for
Quantity
  • Availability
  • Add to Cart