SY3H3D680FCA Synthonix Corporation

Methyl 2-amino-2-methyl-3-phenylpropanoate hydrochloride

  • Cas Number : 64665-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55508.html

Safety Information

MSDS https://synthonix.com/55508.html
Availability details for
Quantity
  • Availability
  • Add to Cart