SY3H3D680FCD Synthonix Corporation

(R)-Methyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride

  • Cas Number : 2049127-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55517.html

Safety Information

MSDS https://synthonix.com/55517.html
Availability details for
Quantity
  • Availability
  • Add to Cart