SY3H3D680FD0 Synthonix Corporation

1-(3-Methoxyphenyl)piperazine hydrochloride

  • Cas Number : 16015-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55520.html

Safety Information

MSDS https://synthonix.com/55520.html
Availability details for
Quantity
  • Availability
  • Add to Cart