SY3H3D680FD1 Synthonix Corporation

1-(2-(Methylsulfonyl)phenyl)piperazine hydrochloride

  • MDL number No MDL found
  • Cas Number : 916488-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55523.html

Safety Information

MSDS https://synthonix.com/55523.html
Availability details for
Quantity
  • Availability
  • Add to Cart