SY3H3D680FD3 Synthonix Corporation

(4-(3-Methoxypropoxy)-3-methylpyridin-2-yl)methanol hydrochloride

  • MDL number No MDL found
  • Cas Number : 675198-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55533.html

Safety Information

MSDS https://synthonix.com/55533.html
Availability details for
Quantity
  • Availability
  • Add to Cart