SY3H3D680FD4 Synthonix Corporation

(2S,4S)-Methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1279039-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55541.html

Safety Information

MSDS https://synthonix.com/55541.html
Availability details for
Quantity
  • Availability
  • Add to Cart