SY3H3D680FD9 Synthonix Corporation

Methyl [2,3'-bipyridine]-6'-carboxylate

  • MDL number No MDL found
  • Cas Number : 845827-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55563.html

Safety Information

MSDS https://synthonix.com/55563.html
Availability details for
Quantity
  • Availability
  • Add to Cart