SY3H3D680FDC Synthonix Corporation

Methyl 4-chloro-2-methylquinoline-8-carboxylate

  • MDL number No MDL found
  • Cas Number : 1234818-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55582.html

Safety Information

MSDS https://synthonix.com/55582.html
Availability details for
Quantity
  • Availability
  • Add to Cart