SY3H3D680FDD Synthonix Corporation

Methyl 5-(4-bromophenyl)-3-methylisoxazole-4-carboxylate

  • Cas Number : 1228689-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55586.html

Safety Information

MSDS https://synthonix.com/55586.html
Availability details for
Quantity
  • Availability
  • Add to Cart