SY3H3D680FE0 Synthonix Corporation

3-Methoxy-2-naphthaldehyde

  • MDL number No MDL found
  • Cas Number : 56679-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/55611.html

Safety Information

MSDS https://synthonix.com/55611.html
Availability details for
Quantity
  • Availability
  • Add to Cart