SY3H3D680FE4 Synthonix Corporation

Methyl 8-hydroxy-1-naphthoate

  • MDL number No MDL found
  • Cas Number : 5247-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55621.html

Safety Information

MSDS https://synthonix.com/55621.html
Availability details for
Quantity
  • Availability
  • Add to Cart