SY3H3D680FE5 Synthonix Corporation

Methyl 2-(2,3-dichlorobenzylidene)-3-oxobutanoate

  • MDL number No MDL found
  • Cas Number : 74073-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55622.html

Safety Information

MSDS https://synthonix.com/55622.html
Availability details for
Quantity
  • Availability
  • Add to Cart