SY3H3D680FE6 Synthonix Corporation

3-(2-Methoxyphenyl)thiophene-2-carboxylic acid

  • Cas Number : 666841-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55623.html

Safety Information

MSDS https://synthonix.com/55623.html
Availability details for
Quantity
  • Availability
  • Add to Cart