SY3H3D680FE9 Synthonix Corporation

4-Methyl-2-phenylpyridine

  • MDL number No MDL found
  • Cas Number : 3475-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55634.html

Safety Information

MSDS https://synthonix.com/55634.html
Availability details for
Quantity
  • Availability
  • Add to Cart