SY3H3D680FEF Synthonix Corporation

5-(4-Methoxyphenyl)pyridin-2(1H)-one

  • MDL number No MDL found
  • Cas Number : 53242-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/55665.html

Safety Information

MSDS https://synthonix.com/55665.html
Availability details for
Quantity
  • Availability
  • Add to Cart