SY3H3D680FF2 Synthonix Corporation

(Z)-2-Methyl-4-(4-methylbenzylidene)oxazol-5(4H)-one

  • Cas Number : 93634-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55671.html

Safety Information

MSDS https://synthonix.com/55671.html
Availability details for
Quantity
  • Availability
  • Add to Cart