SY3H3D680FF4 Synthonix Corporation

Methyl 2-(2-oxo-1,2-dihydroquinolin-3-yl)acetate

  • MDL number No MDL found
  • Cas Number : 156496-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55684.html

Safety Information

MSDS https://synthonix.com/55684.html
Availability details for
Quantity
  • Availability
  • Add to Cart