SY3H3D680FF8 Synthonix Corporation

1-(4-Methoxybenzyl)-1H-pyrazole-4-carbaldehyde

  • MDL number No MDL found
  • Cas Number : 153687-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55716.html

Safety Information

MSDS https://synthonix.com/55716.html
Availability details for
Quantity
  • Availability
  • Add to Cart