SY3H3D680FFA Synthonix Corporation

4-(4-Methoxyphenyl)-2-(methylthio)pyrimidine

  • Cas Number : 148990-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55729.html

Safety Information

MSDS https://synthonix.com/55729.html
Availability details for
Quantity
  • Availability
  • Add to Cart