SY3H3D680FFB Synthonix Corporation

Methyl 2-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1283718-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55751.html

Safety Information

MSDS https://synthonix.com/55751.html
Availability details for
Quantity
  • Availability
  • Add to Cart