SY3H3D680FFC Synthonix Corporation

Methyl 1-((4-fluorophenyl)carbamoyl)cyclopropanecarboxylate

  • MDL number MFCD27939981
  • Cas Number : 1345847-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55755.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55755.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart