SY3H3D680FFD Synthonix Corporation

Methyl 4-acetamido-5-chloro-2,3-dihydrobenzofuran-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 143878-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55756.html

Safety Information

MSDS https://synthonix.com/55756.html
Availability details for
Quantity
  • Availability
  • Add to Cart