SY3H3D680FFE Synthonix Corporation

1-Methoxy-2-methylnaphthalene

  • MDL number No MDL found
  • Cas Number : 14093-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55760.html

Safety Information

MSDS https://synthonix.com/55760.html
Availability details for
Quantity
  • Availability
  • Add to Cart