SY3H3D680FFF Synthonix Corporation

Methyl 1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 7442-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55770.html

Safety Information

MSDS https://synthonix.com/55770.html
Availability details for
Quantity
  • Availability
  • Add to Cart