SY3H3D681000 Synthonix Corporation

Methyl 4-oxo-1,2,3,4-tetrahydronaphthalene-1-carboxylate

  • MDL number No MDL found
  • Cas Number : 156390-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/55771.html

Safety Information

MSDS https://synthonix.com/55771.html
Availability details for
Quantity
  • Availability
  • Add to Cart