SY3H3D681002 Synthonix Corporation

trans-2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid

  • Cas Number : 99226-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55780.html

Safety Information

MSDS https://synthonix.com/55780.html
Availability details for
Quantity
  • Availability
  • Add to Cart