SY3H3D681007 Synthonix Corporation

Methyl 2-(1-methyl-1H-indol-3-yl)acetate

  • Cas Number : 58665-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55819.html

Safety Information

MSDS https://synthonix.com/55819.html
Availability details for
Quantity
  • Availability
  • Add to Cart