SY3H3D68100A Synthonix Corporation

(Z)-Methyl 2-acetamido-3-phenylacrylate

  • MDL number No MDL found
  • Cas Number : 60676-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55826.html

Safety Information

MSDS https://synthonix.com/55826.html
Availability details for
Quantity
  • Availability
  • Add to Cart