SY3H3D68100D Synthonix Corporation

4-Morpholinophenylglyoxal hydrate

  • MDL number No MDL found
  • Cas Number : 852633-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55832.html

Safety Information

MSDS https://synthonix.com/55832.html
Availability details for
Quantity
  • Availability
  • Add to Cart