SY3H3D681010 Synthonix Corporation

Methyl 3-amino-5,6-dimethoxybenzo[b]thiophene-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 198204-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55842.html

Safety Information

MSDS https://synthonix.com/55842.html
Availability details for
Quantity
  • Availability
  • Add to Cart