SY3H3D681013 Synthonix Corporation

2-(5-Methoxy-2-methyl-1H-indol-3-yl)acetamide

  • MDL number No MDL found
  • Cas Number : 15992-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55868.html

Safety Information

MSDS https://synthonix.com/55868.html
Availability details for
Quantity
  • Availability
  • Add to Cart