SY3H3D681015 Synthonix Corporation

1-(Mesitylsulfonyl)-1H-imidazole

  • MDL number MFCD00005284
  • Cas Number : 50257-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/55874.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/55874.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart