SY3H3D681017 Synthonix Corporation

2-(4-Methylpiperazin-1-yl)-5-nitrobenzonitrile

  • MDL number No MDL found
  • Cas Number : 451459-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55892.html

Safety Information

MSDS https://synthonix.com/55892.html
Availability details for
Quantity
  • Availability
  • Add to Cart