SY3H3D68101C Synthonix Corporation

Methyl 4-Benzoylbutyrate

  • Cas Number : 1501-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55924.html

Safety Information

MSDS https://synthonix.com/55924.html
Availability details for
Quantity
  • Availability
  • Add to Cart