SY3H3D68101E Synthonix Corporation

5-Methoxy-2,2-dimethylchroman-4-one

  • MDL number No MDL found
  • Cas Number : 98910-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55931.html

Safety Information

MSDS https://synthonix.com/55931.html
Availability details for
Quantity
  • Availability
  • Add to Cart