SY3H3D68101F Synthonix Corporation

2-(4-Methoxyphenyl)dihydro-2H-pyran-4(3H)-one

  • MDL number No MDL found
  • Cas Number : 1026692-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/55934.html

Safety Information

MSDS https://synthonix.com/55934.html
Availability details for
Quantity
  • Availability
  • Add to Cart