SY3H3D681020 Synthonix Corporation

5-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid

  • Cas Number : 53568-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/55936.html

Safety Information

MSDS https://synthonix.com/55936.html
Availability details for
Quantity
  • Availability
  • Add to Cart