SY3H3D681024 Synthonix Corporation

5-((2-(Methylamino)-5-nitrophenyl)amino)-5-oxopentanoic acid

  • Cas Number : 91644-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/55967.html

Safety Information

MSDS https://synthonix.com/55967.html
Availability details for
Quantity
  • Availability
  • Add to Cart