SY3H3D68102B Synthonix Corporation

Methyl 2-(piperazin-1-yl)benzoate

  • Cas Number : 159974-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56030.html

Safety Information

MSDS https://synthonix.com/56030.html
Availability details for
Quantity
  • Availability
  • Add to Cart