SY3H3D68102F Synthonix Corporation

Methyl 4-(4-methoxyphenyl)butanoate

  • MDL number No MDL found
  • Cas Number : 20637-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56099.html

Safety Information

MSDS https://synthonix.com/56099.html
Availability details for
Quantity
  • Availability
  • Add to Cart