SY3H3D681034 Synthonix Corporation

1-Methyl-4-(4-nitrobenzyl)piperazine

  • MDL number No MDL found
  • Cas Number : 70261-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56122.html

Safety Information

MSDS https://synthonix.com/56122.html
Availability details for
Quantity
  • Availability
  • Add to Cart