SY3H3D681036 Synthonix Corporation

4-(4-Methoxyphenyl)piperazine-1-carboxamide

  • MDL number No MDL found
  • Cas Number : 89026-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/56124.html

Safety Information

MSDS https://synthonix.com/56124.html
Availability details for
Quantity
  • Availability
  • Add to Cart